Merck
  • Hofmann rearrangement of carboxamides mediated by hypervalent iodine species generated in situ from iodobenzene and oxone: reaction scope and limitations.
Hofmann rearrangement of carboxamides mediated by hypervalent iodine species generated in situ from iodobenzene and oxone: reaction scope and limitations.
Organic letters (2010-09-17)
Aleksandra A Zagulyaeva, Christopher T Banek, Mekhman S Yusubov, Viktor V Zhdankin
ZUSAMMENFASSUNG
Alkylcarboxamides can be converted to the respective amines by Hofmann rearrangement using hypervalent iodine species generated in situ from PhI and Oxone in aqueous acetonitrile. On the basis of this reaction, a convenient experimental procedure for the preparation of alkylcarbamates using Oxone as the oxidant in the presence of iodobenzene in methanol has been developed. An efficient method for direct conversion of substituted benzamides to the respective quinone derivatives by treatment with Oxone and iodobenzene in aqueous acetonitrile has also been found.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Iodbenzol, 98%