Merck
  • Carminic acid modified anion exchanger for the removal and preconcentration of Mo(VI) from wastewater.
Carminic acid modified anion exchanger for the removal and preconcentration of Mo(VI) from wastewater.
Journal of hazardous materials (2010-10-15)
Medhat Mohamed El-Moselhy, Arup K Sengupta, Ryan Smith
ZUSAMMENFASSUNG
Removal and preconcentration of Mo(VI) from water and wastewater solutions was investigated using carminic acid modified anion exchanger (IRA743). Various factors influencing the adsorption of Mo(VI), e.g. pH, initial concentration, and coexisting oxyanions were studied. Adsorption reached equilibrium within <10 min and was independent of initial concentration of Mo(VI). Studies were performed at different pH values to find the pH at which maximum adsorption occurred and was determined to be at a pH between 4.0 and 6.0. The Langmuir adsorption capacity (q(max)) was found to be 13.5mg Mo(VI)/g of the adsorbent. The results showed that modification of IRA743 with carminic acid is suitable for the removal of Mo(VI), as molybdate, from water and wastewater samples. The concentration of Mo(VI) was determined spectrophotometrically using bromopyrogallol red as a complexation reagent. This allows the determination of Mo(VI) in the range 1.0-100.0 μg/mL. The obtained material was subjected to efficient regeneration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Karminsäure, ≥90% (HPLC)
Supelco
Karminsäure, analytical standard