Merck
SANS investigation of the photosynthetic machinery of Chloroflexus aurantiacus.
Biophysical journal (2010-10-21)
Kuo-Hsiang Tang, Volker S Urban, Jianzhong Wen, Yueyong Xin, Robert E Blankenship
ZUSAMMENFASSUNG
Green photosynthetic bacteria harvest light and perform photosynthesis in low-light environments, and contain specialized antenna complexes to adapt to this condition. We performed small-angle neutron scattering (SANS) studies to obtain structural information about the photosynthetic apparatus, including the peripheral light-harvesting chlorosome complex, the integral membrane light-harvesting B808-866 complex, and the reaction center (RC) in the thermophilic green phototrophic bacterium Chloroflexus aurantiacus. Using contrast variation in SANS measurements, we found that the B808-866 complex is wrapped around the RC in Cfx. aurantiacus, and the overall size and conformation of the B808-866 complex of Cfx. aurantiacus is roughly comparable to the LH1 antenna complex of the purple bacteria. A similar size of the isolated B808-866 complex was suggested by dynamic light scattering measurements, and a smaller size of the RC of Cfx. aurantiacus compared to the RC of the purple bacteria was observed. Further, our SANS measurements indicate that the chlorosome is a lipid body with a rod-like shape, and that the self-assembly of bacteriochlorophylls, the major component of the chlorosome, is lipid-like. Finally, two populations of chlorosome particles are suggested in our SANS measurements.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethyldodecylamin-N-oxid -Lösung, ~30% in H2O
Sigma-Aldrich
Octyl-β-D-Glucopyranosid, ≥98% (GC)
Sigma-Aldrich
N,N-Dimethyldodecylamin-N-oxid, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Octyl β-D-glucopyranoside solution, ≥95% (HPLC), 50 % (w/v) in H2O
Sigma-Aldrich
Octyl-β-D-Glucopyranosid, BioXtra, ≥98% (GC)