Merck
  • 1,4-addition of lithium diisopropylamide to unsaturated esters: role of rate-limiting deaggregation, autocatalysis, lithium chloride catalysis, and other mixed aggregation effects.
1,4-addition of lithium diisopropylamide to unsaturated esters: role of rate-limiting deaggregation, autocatalysis, lithium chloride catalysis, and other mixed aggregation effects.
Journal of the American Chemical Society (2010-10-22)
Yun Ma, Alexander C Hoepker, Lekha Gupta, Marc F Faggin, David B Collum
ZUSAMMENFASSUNG
Lithium diisopropylamide (LDA) in tetrahydrofuran at -78 °C undergoes 1,4-addition to an unsaturated ester via a rate-limiting deaggregation of LDA dimer followed by a post-rate-limiting reaction with the substrate. Muted autocatalysis is traced to a lithium enolate-mediated deaggregation of the LDA dimer and the intervention of LDA-lithium enolate mixed aggregates displaying higher reactivities than LDA. Striking accelerations are elicited by <1.0 mol % LiCl. Rate and mechanistic studies have revealed that the uncatalyzed and catalyzed pathways funnel through a common monosolvated-monomer-based intermediate. Four distinct classes of mixed aggregation effects are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithiumdiisopropylamid -Lösung, 2.0 M in THF/heptane/ethylbenzene
Sigma-Aldrich
Diisopropylamin, ≥99.5%
Sigma-Aldrich
Lithiumdiisopropylamid, 97%
Sigma-Aldrich
Diisopropylamin, purified by redistillation, 99.95%
Sigma-Aldrich
Diisopropylamin, puriss. p.a., ≥99.0% (GC)
Supelco
Diisopropylamin, analytical standard