Merck
  • A bacterial laccase from marine microbial metagenome exhibiting chloride tolerance and dye decolorization ability.
A bacterial laccase from marine microbial metagenome exhibiting chloride tolerance and dye decolorization ability.
Applied microbiology and biotechnology (2010-10-22)
Zemin Fang, Tongliang Li, Quan Wang, Xuecheng Zhang, Hui Peng, Wei Fang, Yuzhi Hong, Honghua Ge, Yazhong Xiao
ZUSAMMENFASSUNG
Laccases are blue multicopper oxidases with potential applications in environmental and industrial biotechnology. In this study, a new bacterial laccase gene of 1.32 kb was obtained from a marine microbial metagenome of the South China Sea by using a sequence screening strategy. The protein (named as Lac15) of 439 amino acids encoded by the gene contains three conserved Cu(2+)-binding domains, but shares less than 40% of sequence identities with all of the bacterial multicopper oxidases characterized. Lac15, recombinantly expressed in Escherichia coli, showed high activity towards syringaldazine at pH 6.5-9.0 with an optimum pH of 7.5 and with the highest activity occurring at 45 °C. Lac15 was stable at pH ranging from 5.5 to 9.0 and at temperatures from 15 to 45 °C. Distinguished from fungal laccases, the activity of Lac15 was enhanced twofold by chloride at concentrations lower than 700 mM, and kept the original level even at 1,000 mM chloride. Furthermore, Lac15 showed an ability to decolorize several industrial dyes of reactive azo class under alkalescent conditions. The properties of alkalescence-dependent activity, high chloride tolerance, and dye decolorization ability make the new laccase Lac15 an alternative for specific industrial applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Syringaldazin, indicator for laccase and peroxidase activity
Sigma-Aldrich
Syringaldazin, 98%