Merck
  • Facile conversion of tetracycline antibiotics to 4,11a-bridged derivatives via oxidative mannich cyclization.
Facile conversion of tetracycline antibiotics to 4,11a-bridged derivatives via oxidative mannich cyclization.
The Journal of antibiotics (2010-10-28)
Jianxin Gu, Ping Cai, Yumin Gong, Mark E Ruppen, Thomas Storz
ZUSAMMENFASSUNG
Treatment of a variety of tetracyclines (tigecycline, minocycline, tetracycline and doxycycline) with Ag(2)CO(3)/EDTA or Hg(OAc)(2) cleanly gave the 4,11a-bridged derivatives in high yields. The reactions proceeded through a novel, intramolecular Mannich cyclization of an iminium species generated by oxidation of the tertiary dimethylamino group at C(4) by Ag(I) or Hg(II). Tetracyclines without 5-OH-substitution (tigecycline, tetracycline and minocycline) gave the 4-OH-substituted, 4,11a-bridged compound, whereas doxycycline gave the 4-dimethylamino-substituted, 4,11a-bridged product. In the case of tetracycline, the 4,11a-bridged compound can equilibrate further to a 4,6-bridged hemiketal. Some of the bridged compounds underwent a novel decarboxylation--rearrangement sequence under acidic conditions to give tricyclic, open chain 1,4-quinoid compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Quecksilber(II)-acetat, ACS reagent, ≥98.0%
Sigma-Aldrich
Silbercarbonat, 99%
Sigma-Aldrich
Silbercarbonat auf Celit®, extent of labeling: ~50 wt. % loading
Sigma-Aldrich
Silbercarbonat, purum p.a., ≥99.0% (AT)