Merck
  • Susceptibility of Varroa destructor (Acari: Varroidae) to synthetic acaricides in Uruguay: Varroa mites' potential to develop acaricide resistance.
Susceptibility of Varroa destructor (Acari: Varroidae) to synthetic acaricides in Uruguay: Varroa mites' potential to develop acaricide resistance.
Parasitology research (2010-10-28)
Matías Daniel Maggi, Sergio Roberto Ruffinengo, Yamandú Mendoza, Pilar Ojeda, Gustavo Ramallo, Iganazio Floris, Martín Javier Eguaras
ZUSAMMENFASSUNG
The purpose of this study was to estimate the acaricide susceptibility of Varroa destructor populations from Uruguay, which had never been exposed to synthetic acaricides. It was also to determine whether acaricide resistance to coumaphos occurred in apiaries in which acaricide rotation had been applied. Bioassays with acaricides against mite populations that had never been exposed to synthetic acaricides were performed, also against mite populations in which control failures with coumaphos had been reported. Additionally, coumaphos' effectiveness in honeybee colonies was experimentally tested. The lethal concentration that kills 50% of the exposed animals (LC(50)) for susceptible mite populations amounted to 0.15 μg/Petri dish for coumaphos and to less than 0.3 μg/Petri dish for the other acaricides. Coumaphos LC(50) was above 40 μg/Petri dish for resistant mites. The effectiveness of coumaphos in honeybee colonies parasitized by V. destructor ranged from 17.6% to 93.9%. LC(50) for mite populations susceptible to the most commonly applied miticides was determined, and the first case of coumaphos resistance recorded in Uruguay was established.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Coumaphos, PESTANAL®, analytical standard