Merck
  • Synthesis and characterization of amphiphilic glycidol-chitosan-deoxycholic acid nanoparticles as a drug carrier for doxorubicin.
Synthesis and characterization of amphiphilic glycidol-chitosan-deoxycholic acid nanoparticles as a drug carrier for doxorubicin.
Biomacromolecules (2010-10-30)
Huofei Zhou, Weiting Yu, Xin Guo, Xiudong Liu, Nan Li, Ying Zhang, Xiaojun Ma
ZUSAMMENFASSUNG
Novel amphiphilic chitosan derivatives (glycidol-chitosan-deoxycholic acid, G-CS-DCA) were synthesized by grafting hydrophobic moieties, deoxycholic acid (DCA), and hydrophilic moieties, glycidol, with the purpose of preparing carriers for poorly soluble drugs. Based on self-assembly, G-CS-DCA can form nanoparticles with size ranging from 160 to 210 nm, and G-CS-DCA nanoparticles maintained stable structure for about 3 months when stored in PBS (pH 7.4) at room temperature. The critical aggregation concentration decreased from 0.043 mg/mL to 0.013 mg/mL with the increase of degree of substitution (DS) of DCA. Doxorubicin (DOX) could be easily encapsulated into G-CS-DCA nanoparticles and keep a sustained release manner without burst release when exposed to PBS (pH 7.4) at 37 °C. Antitumor efficacy results showed that DOX-G-CS-DCA have significant antitumor activity when MCF-7 cells were incubated with different concentration of DOX-G-CS-DCA nanoparticles. The fluorescence imaging results indicated DOX-G-CS-DCA nanoparticles could easily be uptaken by MCF-7 cells. These results suggested that G-CS-DCA nanoparticles may be a promising carrier for DOX delivery in cancer therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glyceringlycid, 96%
Sigma-Aldrich
Glycidol chitosan, Degree of functionalization > 80%