Merck
Fused bicyclic derivatives of 2,4-diaminopyrimidine as c-Met inhibitors.
Bioorganic & medicinal chemistry letters (2010-12-03)
Linda R Weinberg, Mark S Albom, Thelma S Angeles, Jean Husten, Joseph G Lisko, Robert J McHugh, Karen L Milkiewicz, Seetha Murthy, Gregory R Ott, Jay P Theroff, Rabindranath Tripathy, Ted L Underiner, Craig A Zificsak, Bruce D Dorsey
ZUSAMMENFASSUNG
The HGF-c-Met signaling axis is an important paracrine mediator of epithelial-mesenchymal cell interactions involving the regulation of multiple cellular activities including cell motility, mitogenesis, morphogenesis, and angiogenesis. Dysregulation of c-Met signaling (e.g., overexpression or increased activation) is associated with the development of a wide range of tumor types; thus, inhibiting the HGF-c-Met pathway is predicted to lead to anti-tumor effects in many cancers. Elaboration of a 2-arylaminopyrimidine scaffold led to a series of potent c-Met inhibitors bearing a C4-2-amino-N-methylbenzamide group. Specifically, a series of C2-benzazepinone analogs demonstrated potent inhibition of c-Met in enzymatic and cellular assays. Kinase selectivity could be tuned by varying the nature of the alkyl group on the benzazepinone nitrogen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,4-Diaminopyrimidin, 98%