Merck
  • The size control of silver nanocrystals with different polyols and its application to low-reflection coating materials.
The size control of silver nanocrystals with different polyols and its application to low-reflection coating materials.
Nanotechnology (2010-12-16)
Keum Hwan Park, Sang Hyuk Im, O Ok Park
ZUSAMMENFASSUNG
The size of silver nanocrystals in polyol synthesis can be simply controlled by tuning the viscosity of the reaction medium such as ethylene glycol, 1,2-propanediol, 1,4-butanediol and 1,5-pentanediol. We found that a higher viscose medium (1,5-pentanediol) led to monodispersed smaller particles thanks to the slow addition of silver atoms into the nuclei. Size-controlled silver nanocrystals of 30 nm were obtained in a viscosity controlled medium of 1,5-pentanediol to synthesize a low refractive index filler by coating with silica and subsequent etching of the silver core. The coated low-reflection layer from the hollow silica nanoparticles on polyethylene terephthalate (PET) film can greatly reduce the reflection of the PET film from 10% to 2% over the entire visible region.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,5-Pentandiol, 96%
Sigma-Aldrich
1,5-Pentandiol, purum, ≥97.0% (GC)