Merck
Biotransformation of turmerones by Aspergillus niger.
Journal of natural products (2010-12-30)
Mai Fujiwara, Shinsuke Marumoto, Nobuo Yagi, Mitsuo Miyazawa
ZUSAMMENFASSUNG
Biotransformation studies conducted on (+)-(S)-ar-turmerone (1) and (+)-(S)-dihydro-ar-turmerone (2) by the fungus Aspergillus niger have revealed that 1 was metabolized to give four oxidized metabolites, (+)-(7S)-hydroxydehydro-ar-todomatuic acid (3), (+)-(7S,10E)-12-hydroxydehydro-ar-todomatuic acid (4), (+)-(7S,10E)-7,12-dihydroxydehydro-ar-todomatuic acid (5), and (+)-(7S)-15-carboxy-9,13-epoxy-7-hydroxy-9,13-dehydro-ar-curcumene (6), and (+)-(S)-dihydro-ar-turmerone (2) was metabolized to (+)-7,11-dihydroxy-ar-todomatuic acid (7). Metabolites 3-7 were characterized using spectroscopic techniques. Metabolites 3-7 inhibited acetylcholinesterase (AChE) although less so than the parent substrates.