Merck
  • A novel α-isocyanoacetamide-based three-component reaction for the synthesis of dialkyl 2-acyl-5-aminofuran-3,4-dicarboxylates.
A novel α-isocyanoacetamide-based three-component reaction for the synthesis of dialkyl 2-acyl-5-aminofuran-3,4-dicarboxylates.
Organic & biomolecular chemistry (2011-01-21)
Riccardo Mossetti, Diego Caprioglio, Giampiero Colombano, Gian Cesare Tron, Tracey Pirali
ZUSAMMENFASSUNG
α-Isocyanoacetamides, acyl chlorides and dialkylacetylenedicarboxylates undergo a smooth multicomponent reaction to produce dialkyl 2-acyl-5-aminofuran-3,4-dicarboxylates in good yield. The scope and mechanism of this new multicomponent transformation are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyanoacetamid, 99%