Merck
  • Spectroscopic and computational characterization of CuII-OOR (R = H or cumyl) complexes bearing a Me6-tren ligand.
Spectroscopic and computational characterization of CuII-OOR (R = H or cumyl) complexes bearing a Me6-tren ligand.
Dalton transactions (Cambridge, England : 2003) (2011-01-25)
Yu Jin Choi, Kyung-Bin Cho, Minoru Kubo, Takashi Ogura, Kenneth D Karlin, Jaeheung Cho, Wonwoo Nam
ZUSAMMENFASSUNG
A copper(II)-hydroperoxo complex, [Cu(Me(6)-tren)(OOH)](+) (2), and a copper(ii)-cumylperoxo complex, [Cu(Me(6)-tren)(OOC(CH(3))(2)Ph)](+) (3), were synthesized by reacting [Cu(Me(6)-tren)(CH(3)CN)](2+) (1) with H(2)O(2) and cumyl-OOH, respectively, in the presence of triethylamine. These intermediates, 2 and 3, were successfully characterized by various physicochemical methods such as UV-vis, ESI-MS, resonance Raman and EPR spectroscopies, leading us to propose structures of the Cu(II)-OOR species with a trigonal-bipyramidal geometry. Density functional theory (DFT) calculations provided geometric and electronic configurations of 2 and 3, showing trigonal bipyramidal copper(II)-OOR geometries. These copper(II)-hydroperoxo and -cumylperoxo complexes were inactive in electrophilic and nucleophilic oxidation reactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cumolhydroperoxid, technical grade, 80%