Merck
  • Effects of Piriformospora indica and Sebacina vermifera on growth and yield of essential oil in fennel (Foeniculum vulgare) under greenhouse conditions.
Effects of Piriformospora indica and Sebacina vermifera on growth and yield of essential oil in fennel (Foeniculum vulgare) under greenhouse conditions.
Journal of basic microbiology (2011-01-25)
Hossein Kari Dolatabadi, Ebrahim Mohammadi Goltapeh, Kamkar Jaimand, Neda Rohani, Ajit Varma
ZUSAMMENFASSUNG
Fennel (Foeniculum vulgare) is a very important plant in the family of Apiaceae. Effects of inoculation of two endophytic fungi (Piriformospora indica and Sebacina vermifera) in growth, yield and composition of the essential oil of fennel (F. vulgare) were evaluated in pot cultures. Dry fruits were ground with an electric grinder and oil was extracted by hydrodistillation, and their composition was determined by GC/MS. In pot experiment, the maximum dry weight of the green tissue and root and plant height were obtained with P. indica, and maximum number of umbels per plant and dry weight of 1000 fruits were produced with S. vermifera. The P. indica and S. vermifera inoculation significantly increased oil yield as compared to non-inoculated control plants. GC and GC/MS studies revealed that the level of anethole was increased with P. indica and S. vermifera.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
trans-Anethol, 99%
Sigma-Aldrich
trans-Anethol, ≥99%, FCC, FG
Supelco
trans-Anethol, analytical standard
Supelco
trans-Anethol, Pharmaceutical Secondary Standard; Certified Reference Material