Merck
Protective effects of sodium molybdate on carbon tetrachloride-induced hepatotoxicity in rats.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2011-02-01)
Akram Eidi, Maryam Eidi, Mahsa Al-Ebrahim, Ali Haeri Rohani, Pejman Mortazavi
ZUSAMMENFASSUNG
Molybdenum is an essential trace micronutrient element that plays an important role in animal and plant physiology. Molybdenum is a constituent of at least three mammalian metalloflavoproteins: xanthine oxidase, aldehyde oxidase and sulphite oxidase. In the present study, the hepatoprotective potential of sodium molybdate was investigated against carbon tetrachloride (CCl(4))-induced liver damage in rats. Administration of CCl(4) increased the serum alanine aminotransferase, aspartate aminotransferase and alkaline phosphatase levels in rats and reduced levels of the antioxidant enzymes superoxide dismutase and catalase in the liver. Treatment with sodium molybdate significantly attenuated these changes to nearly undetectable levels. The histopathological changes induced by CCl(4) were also significantly attenuated by sodium molybdate treatment. Therefore, the results of this study suggest that sodium molybdate can protect the liver against CCl(4)-induced oxidative damage in rats, and this hepatoprotective effect might be attributable to its modulation of detoxification enzymes and/or its antioxidant and free radical scavenger effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriummolybdat Dihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Natriummolybdat Dihydrat, ≥99.5%
Sigma-Aldrich
Natriummolybdat, ≥98%
Sigma-Aldrich
Natriummolybdat Dihydrat, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
Natriummolybdat, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Natriummolybdat Dihydrat, 99.99% trace metals basis