Merck
Evolution of fat crystal network microstructure followed by NMR.
Journal of agricultural and food chemistry (2011-02-15)
Matthieu Adam-Berret, Marine Boulard, Alain Riaublanc, François Mariette
ZUSAMMENFASSUNG
Model systems composed of tristearin in solid state and tricaprin in liquid state with different solid-fat content (SFC) and storage time have been investigated by relaxation NMR and NMR diffusometry. The T(2) relaxation of the tricaprin in the melt exhibited a bimodal distribution as previously observed. The SFC had a major effect on the T(2) relaxation. This effect was explained according to the fast diffusive exchange model in porous media. According to this model the changes in T(2) relaxation as a function of the SFC and storage time were explained by the decrease of the surface-to-volume ratio of the crystal induced by Ostwald ripening. The diffusion coefficient D of the tricaprin in the melt decreased for higher SFC. Since no significant variation of D was observed for different diffusion time, D reflected the long-range connectivity and the tortuosity was calculated. During storage the diffusion coefficient remained constant.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin-tricaprinat, ≥99% (GC)
Sigma-Aldrich
Glycerintristearat, ≥99%
Sigma-Aldrich
Glycerintristearat, technical