Merck
  • Impact of a heteroatom in a structure-activity relationship study on analogues of phenyl glycidyl ether (PGE) from epoxy resin systems.
Impact of a heteroatom in a structure-activity relationship study on analogues of phenyl glycidyl ether (PGE) from epoxy resin systems.
Chemical research in toxicology (2011-03-05)
Ida B Niklasson, Tamara Delaine, Kristina Luthman, Ann-Therese Karlberg
ZUSAMMENFASSUNG
Epoxy resins are among the most common causes of occupational contact dermatitis. They are normally used in so-called epoxy resin systems (ERS). These commercial products are combinations of epoxy resins, curing agents, modifiers, and reactive diluents. The most frequently used resins are diglycidyl ethers based on bisphenol A (DGEBA) and bisphenol F (DGEBF). In this study, we have investigated the contact allergenic properties of a series of analogues to the reactive diluent phenyl glycidyl ether (PGE), all with similar basic structures but with varying heteroatoms or with no heteroatom present. The chemical reactivity of the compounds in the test series toward the hexapeptide H-Pro-His-Cys-Lys-Arg-Met-OH was investigated. All epoxides were shown to bind covalently to both cysteine and proline residues. The percent depletion of nonreacted peptide was also studied resulting in ca. 60% depletion when using either PGE, phenyl 2,3-epoxypropyl sulfide (2), or N-(2,3-epoxypropyl)aniline (3), and only 15% when using 1,2-epoxy-4-phenylbutane (4) at the same time point. The skin sensitization potencies of the epoxides using the murine local lymph node assay (LLNA) were evaluated in relation to the observed physicochemical and reactivity properties. To enable determination of statistical significance between structurally closely related compounds, a nonpooled LLNA was performed. It was found that all investigated compounds containing a heteroatom in the α-position to the epoxide were strong sensitizers, congruent with the reactivity data, indicating that the impact of a heteroatom is crucial for the sensitizing capacity for this type of epoxides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenyl-glycidether, 99%
Sigma-Aldrich
Phenyl-glycidether, technical grade