Merck
  • Protective effect of photodegradation product of nifedipine against tumor necrosis factor alpha-induced oxidative stress in human glomerular endothelial cells.
Protective effect of photodegradation product of nifedipine against tumor necrosis factor alpha-induced oxidative stress in human glomerular endothelial cells.
The journal of medical investigation : JMI (2011-03-05)
Yayoi Fukuhara, Koichiro Tsuchiya, Yuya Horinouchi, Soichiro Tajima, Yoshitaka Kihira, Shuichi Hamano, Kazuyoshi Kawazoe, Yasumasa Ikeda, Keisuke Ishizawa, Shuhei Tomita, Toshiaki Tamaki
ZUSAMMENFASSUNG
Recently, increasing evidence suggests that the antihypertensive drug nifedipine acts as a protective agent for endothelial cells, and that the activity is unrelated to its calcium channel blocking. Nitrosonifedipine (NO-NIF) is metabolically and photochemically produced from nifedipine, and NO-NIF has been recognized as a contaminant of nifedipine because it has no antihypertensive effect. Treatment of tumor necrosis factor-α (TNF-α) suppressed the cell viability and facilitated the expression of Inter-Cellular Adhesion Molecule 1(ICAM-1) in human glomerular endothelial cells (HGECs) though, pretreatment of NO-NIF significantly recovered the TNF-α-induced cell damage to the same extent as Trolox-C did, and suppressed the ICAM-1 expression in a concentration dependent manner. In addition, NO-NIF inhibited the cell toxicity induced by cumene hydroperoxide, which hampers the integrity of cell membrane through oxidative stress, as effective as Trolox-c. These data suggest that NO-NIF is a candidate for a new class of antioxidative drug that protect cells against oxidative stress in glomerular endothelial cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cumolhydroperoxid, technical grade, 80%