Merck
  • Selective oxidation of benzylic and allylic alcohols using Mn(OAc)3/catalytic 2,3-dichloro-5,6-dicyano-1,4-benzoquinone.
Selective oxidation of benzylic and allylic alcohols using Mn(OAc)3/catalytic 2,3-dichloro-5,6-dicyano-1,4-benzoquinone.
Organic letters (2011-03-15)
Casey C Cosner, Pablo J Cabrera, Katherine M Byrd, Asia M Adams Thomas, Paul Helquist
ZUSAMMENFASSUNG
A practical, chemoselective oxidation of alcohols employing catalytic quantities of DDQ as the oxidant and Mn(OAc)(3) as the co-oxidant is described. Electron-rich benzylic alcohols are oxidized efficiently to their corresponding carbonyls, but less electron-rich benzylic alcohols remain unchanged. Allylic alcohols are rapidly oxidized to their corresponding aldehyde or ketone counterparts in high yields. This protocol is operationally simple, employs an inexpensive source of Mn(OAc)(3), has short reaction times, and exhibits a significant chemoselectivity favoring allylic alcohols over benzylic alcohols. This procedure also avoids the use of very large excesses of reagents and sometimes poor reproducibility that characterize previously developed reagents such as MnO(2).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,3-Dichlor-5,6-dicyano-p-benzochinon, 98%