Merck
Analysis of the essential oil of Amsonia illustris.
Natural product communications (2011-03-24)
Annamaria London, Katalin Veres, Krisztina Szabó, Erzsébet Háznagy-Radnai, Imre Máthé
ZUSAMMENFASSUNG
The root and leaf essential oils, present in trace amounts in Amsonia illustris Woods. (Apocynaceae), were isolated by steam distillation and their chemical constituents identified by GC-FID and GC-MS. More than 80% of the thirty volatile compounds in the leaf oil were identified, the major constituents being mainly sesquiterpenes like a-humulene (14.5%), beta-caryophyllene (12.4%) and guaiol (11.6%). The volatile ingredients of the root oil were pinocampheol, methyl salicylate, (2E,4E)- decadienal, eugenol and trans-isoeugenol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Supelco
Isoeugenol, analytical standard
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG