Merck
Telomerase-mediated apoptosis of chicken lymphoblastoid tumor cell line by lanthanum chloride.
Biological trace element research (2011-03-31)
Ziwei Zhang, Jintao Wang, Jinlong Li, Shiwen Xu
ZUSAMMENFASSUNG
To investigate the biological effects of lanthanum on chicken lymphoid tumors, cultures of the MDCC-MSB1 chicken lymphoblastoid cell line were treated with 2.5 mM lanthanum chloride for 2 days at 12-h intervals. The apoptotic cells were detected using the TdT-mediated dUTP nick end labeling assay and flow cytometer analysis. The telomerase activity and the chTERT mRNA expression levels of the MDCC-MSB1 cells were examined. The results showed that MDCC-MSB1 cell apoptosis occurred after incubation for 12 to 48 h induced by 2.5 mM LaCl(3). Consistent with the apoptosis results, telomerase activities in LaCl(3)-treated cells significantly decreased (P < 0.05) compared with 0 h group. Furthermore, the transcription of chTERT gene in MDCC-MSB1 cell was significantly inhibited in LaCl(3) treatment group (P < 0.05). These results suggest that the decrease of the chTERT transcription and telomerase activity play an important role in the La-induced apoptosis in chicken lymphoid tumor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lanthan(III)-chlorid Heptahydrat, ACS reagent
Sigma-Aldrich
Lanthan(III)-chlorid Heptahydrat, 99.999% trace metals basis
Sigma-Aldrich
Lanthan(III)-chlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Lanthan(III)-chlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis