Merck
  • Electrochemical impedance determination of polychlorinated biphenyl using a pyrenecyclodextrin-decorated single-walled carbon nanotube hybrid.
Electrochemical impedance determination of polychlorinated biphenyl using a pyrenecyclodextrin-decorated single-walled carbon nanotube hybrid.
Chemical communications (Cambridge, England) (2011-04-01)
Yan Wei, Ling-Tao Kong, Ran Yang, Lun Wang, Jin-Huai Liu, Xing-Jiu Huang
ZUSAMMENFASSUNG
This work reports the first detailed study on an electrochemical impedance sensor for determination of polychlorinated biphenyl (PCB), such as 3,3',4,4'-tetrachlorobiphenyl (PCB-77), based on a single-walled carbon nanotube/pyrenecyclodextrin (SWCNT/PyCD) hybrid.