Merck
  • Conformational consequences of cooperative binding of a coiled-coil peptide motif to poly(N-(2-hydroxypropyl) methacrylamide) HPMA copolymers.
Conformational consequences of cooperative binding of a coiled-coil peptide motif to poly(N-(2-hydroxypropyl) methacrylamide) HPMA copolymers.
Journal of controlled release : official journal of the Controlled Release Society (2011-04-05)
Peter C Griffiths, Alison Paul, Bojana Apostolovic, Harm-Anton Klok, Edoardo de Luca, Stephen M King, Richard K Heenan
ZUSAMMENFASSUNG
Small-angle neutron scattering and pulsed-gradient spin-echo NMR have been used to examine the solution conformation of a series of water soluble poly(N-(2-hydroxypropyl) methacrylamide) P(HPMA) co-polymer drug delivery vehicles incorporating a coiled-coil peptide motif as a novel pH sensitive non-covalent linker. The conformation of the HPMA homopolymer is well-described by a Gaussian coil model and changing pH from pH 7 to pH 5 has little effect on the solution conformation, as quantified via the radius of gyration. Copolymerisation with 5-10mol% of the K3 peptide bearing methacrylate monomer (K3-MA), gave a series of copolymers that exhibited an increase in radius of gyration at both pH's, despite being typically 30% lower in molecular weight, indicating that the K3-MA causes a perturbation (expansion) of the copolymer conformation. Subsequent addition of an equimolar amount of the complementary peptide E3 makes little further difference to the conformation, indicative of the intimate binding (coiled-coil motif) between the two peptides. Again, the effects of pH are small. Only the addition of a large aromatic structure such as methotrexate causes a further perturbation of the structure - the hydrophobic interaction between the MTX units causes a significant collapse of the polymer coil. These findings further elaborate the understanding of those factors that determine the solution conformation of novel polymer therapeutics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydroxypropylmethacrylat, Mixture of hydroxypropyl and hydroxyisopropyl methacrylates, 97%, contains 180-220 ppm monomethyl ether hydroquinone as inhibitor