Merck
Dequalinium induces human leukemia cell death by affecting the redox balance.
Leukemia research (2011-04-12)
Ana I García-Pérez, Eva Galeano, Elena Nieto, Pilar Sancho
ZUSAMMENFASSUNG
Dequalinium, an amphiphilic quinolinium derivative, selectively accumulates in mitochondria and displays anticancer activity in cells from different malignancies. Previous studies indicate a differential DQA-induced cytotoxicity in NB4 and K562 human leukemia cells as a consequence of an early disturbance in mitochondrial function. Results in this paper show that DQA induces a concentration-dependent oxidative stress by decreasing GSH level and increasing ROS in a cell type specific way. Inhibitors of the JNK and p38 stress regulated kinases potentiate DQA-induced NB4 cell death suggesting a protective function for these enzymes. K562 cells with relatively high GSH levels remained resistant to DQA action.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Dequaliniumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material