Merck
Application of alkyne-TCNQ addition reaction to polymerization.
Macromolecular rapid communications (2011-04-12)
Yusuke Washino, Tsuyoshi Michinobu
ZUSAMMENFASSUNG
The polymerization using a high-yielding addition reaction between electron-rich alkynes and 7,7,8,8-tetracyanoquinodimethane (TCNQ) derivatives is described. The bifunctional monomer containing two TCNQ moieties and the counter comonomer bearing two dialkylaniline (DAA)-substituted alkynes are reacted in 1,2-dichloroethane under mild heating conditions. At the high monomer concentrations, high molecular weight linear polymers are obtained, while the reaction at the low monomer concentrations produces a significant amount of the cyclic compounds. A clear relationship between the monomer concentration and the cyclic compound amount is demonstrated. The obtained polymers feature a sufficient thermal stability with the decomposition temperature exceeding 300  °C as well as strong charge-transfer (CT) bands and redox activities ascribed to the produced donor-acceptor moieties. These features are also used to optimize the polymerization conditions and to estimate the chemical structures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
7,7,8,8-Tetracyanochinodimethan, 98%