Merck
  • Simultaneous polarographic determination of 2-nitrophenol and 4-nitrophenol by differential pulse polarography method and support vector regression.
Simultaneous polarographic determination of 2-nitrophenol and 4-nitrophenol by differential pulse polarography method and support vector regression.
Environmental monitoring and assessment (2011-04-13)
Karim Asadpour-Zeynali, Payam Soheili-Azad
ZUSAMMENFASSUNG
A differential pulse polarography (DPP) for the simultaneous determination of 2-nitrophenol and 4-nitrophenol was proposed. It was found that under optimum experimental conditions (pH = 5, scan rate = 5 mV/s, pulse amplitude = -50 mV), 2-nitrophenol and 4-nitrophenol had well-defined polarographic reduction waves with peak potentials at -317 and -406 mV, respectively. In the mixture of two compounds overlapping polarographic peaks were observed. In this study, support vector regression (SVR) was applied to resolve the overlapped polarograms. Furthermore, a comparison was made between the performance of SVR and partial least square (PLS) on data set. The results demonstrated that SVR is a better well-performing alternative for the analysis and modeling of DPP data than the commonly applied PLS technique. The proposed method was used for the determination of 2-nitrophenol and 4-nitrophenol in industrial waste water.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Nitrophenol, 98%