Merck
Persulfate oxidation for the aniline degradation in aqueous systems.
Water science and technology : a journal of the International Association on Water Pollution Research (2011-04-22)
Jin Anotai, Nalinrut Masomboon, Chung-Lin Chuang, Ming-Chun Lu
ZUSAMMENFASSUNG
Ferrous catalyzed persulfate oxidation of dissolved aniline was investigated in aqueous systems under a variety of ferrous ion concentrations and temperature. Result showed that the addition of ferrous ions accelerated the degradation of aniline by persulfate. For the thermally activated persulfate oxidation experiment, the optimum persulfate/aniline concentration ratio at 30˚C was at 5.4 mM or 20/1. This ratio gave the highest aniline removal of 45%. For the ferrous ion catalyzed persulfate oxidation experiment, there was marginal difference in the result for the various ferrous ion/oxidant molar ratios. Thus, another series of experiment was conducted to determine the optimum ratio and a ferrous ion/persulfate molar ratio of 1.25/1 showed the highest removal efficiency.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumpersulfat, reagent grade, ≥98%
Sigma-Aldrich
Natriumpersulfat, BioUltra, ≥99%
Sigma-Aldrich
Natriumpersulfat, BioXtra, ≥99%