Merck
Production of biodiesel and lactic acid from rapeseed oil using sodium silicate as catalyst.
Bioresource technology (2011-05-03)
Yun-Duo Long, Feng Guo, Zhen Fang, Xiao-Fei Tian, Li-Qun Jiang, Fan Zhang
ZUSAMMENFASSUNG
Biodiesel and lactic acid from rapeseed oil was produced using sodium silicate as catalyst. The transesterification in the presence of the catalyst proceeded with a maximum yield of 99.6% under optimized conditions [3% (w/w) sodium silicate, methanol/oil molar ratio 9/1, reaction time 60 min, reaction temperature 60°C, and stirring rate 250 rpm]. After six consecutive transesterification reactions, the catalyst was collected and used for catalysis of the conversion of glycerol to lactic acid. A maximum yield of 80.5% was achieved when the reaction was carried out at a temperature of 300°C for 90 min. Thus, sodium silicate is an effective catalyst for transesterification and lactic acid production from the biodiesel by-product, glycerol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriummetasilicat Pentahydrat, ≥95.0% (T)