Merck
  • Simultaneous determination of aflatoxins B₁, B₂, G₁, and G₂ in foods and feed materials.
Simultaneous determination of aflatoxins B₁, B₂, G₁, and G₂ in foods and feed materials.
Methods in molecular biology (Clifton, N.J.) (2011-05-14)
Marilena Muscarella, Marco Iammarino, Donatella Nardiello, Sonia Lo Magro, Carmen Palermo, Diego Centonze
ZUSAMMENFASSUNG
A high-performance liquid chromatographic method with on-line post-column photochemical derivatization and fluorimetric detection for the simultaneous separation and quantitative determination of aflatoxin (AF) B(1), B(2), G(1), and G(2) in foodstuffs and feed materials is reported.The chromatographic separation is accomplished by using a C(18) column eluted with an isocratic mobile phase consisting of water, methanol, and acetonitrile. The sample preparation requires a simple extraction of aflatoxins with a mixture of water and methanol, and a purification step by immunoaffinity column clean-up. The total analysis time, including sample preparation and chromatographic separation, does not exceed 40 min with a run time of 10 min. The procedure for the determination of aflatoxins in food samples and cereals for animal consumption has been extensively validated, in agreement with Regulation (EC) No. 882/2004, demonstrating the conformity of the method with provisions of Regulation (EC) No. 401/2006 in terms of sensitivity, linearity, selectivity, and precision.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aflatoxin G2
Sigma-Aldrich
Aflatoxin B2
Sigma-Aldrich
Aflatoxin G1 aus Aspergillus flavus, from Aspergillus flavus
Supelco
Aflatoxin B2 -Lösung, 0.5 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin G2 -Lösung, 0.5 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin G1 -Lösung, 2 μg/mL in acetonitrile, analytical standard
Aflatoxin G2 -Lösung, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Aflatoxin B2 -Lösung, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Aflatoxin G1 -Lösung, 3.78 μg/g in acetonitrile, ERM®, certified reference material