Merck
  • [Simultaneous determination of five yellow dyes in foods by high performance liquid chromatography coupled with tandem mass spectrometry].
[Simultaneous determination of five yellow dyes in foods by high performance liquid chromatography coupled with tandem mass spectrometry].
Se pu = Chinese journal of chromatography (2011-05-18)
Saijun Lin, Haiyun Tu, Lan Sun, Hailong Xiao, Xiangrong Pan, Xiaoyan Ma
ZUSAMMENFASSUNG
A method based on high performance liquid chromatography coupled with tandem mass spectrometry (HPLC-MS/MS) was developed for simultaneous determination of five yellow industrial dyes. The separations were performed with an Agilent ODS C18 column at a flow rate of 0.3 mL/min. The mobile phase was 5 mmol/L ammonium acetate (containing 0.1% formic acid)-acetonitrile (3:2, v/v). Under the optimized detection conditions, the linear ranges for Chrysoidine G and Basic Yellow 2 were 5.0-80.0 mg/L, and for Acid Orange I, Acid orange Il and Acid Yellow 36 were 10.0-160.0 microg/L. The limits of quantification for Chrysoidine G, Basic Yellow 2, Acid Orange I, Acid Orange II and Acid Yellow 36 were 20, 20, 40, 40 and 40 ng/g, respectively. The relative standard deviations of reproducibility of this method for retention time and peak area were no more than 0.50% and 2.14%, respectively. The method was applied to determine the recoveries of the above five dyes in chicken, bean products and yellow croaker were between 79.8%-95.2% with satisfactory results.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Metanilgelb, Dye content 70 %
Supelco
Metanilgelb, analytical standard