Merck
Solvent and TMEDA effects on the configurational stability of chiral lithiated aryloxiranes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-05-28)
Filippo Maria Perna, Antonio Salomone, Mariangela Dammacco, Saverio Florio, Vito Capriati
ZUSAMMENFASSUNG
The employment of hexane/N,N,N',N'-tetramethylethylenediamine (TMEDA) dramatically hinders the racemization of those lithiated styrene oxides (trifluoromethyl-, chloro-, and phenylthio-substituted) that have been proven to be configurationally unstable in THF on the timescale of their reactions. The barriers to inversion and the activation parameters, calculated (Eyring equation) for reactions performed in THF, THF/TMEDA, and hexane/TMEDA, suggest the intervention of particular enantiomerization mechanisms for each case. The role of TMEDA in both coordinating and noncoordinating solvents has also been questioned and discussed in light of the kinetic data gathered and a model for deprotonation in hexane/TMEDA has also been proposed. The synthetic benefits of our results became apparent on establishing an asymmetric synthesis of an industrially important antifungal agent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
Phenylethylenoxid, 97%
Sigma-Aldrich
(S)-Phenylethylenoxid, 98%, optical purity ee: 98% (GC)
Sigma-Aldrich
(R)-Phenylethylenoxid, 97%, optical purity ee: 97% (GLC)
Sigma-Aldrich
(R)-Phenylethylenoxid, ChiPros®, produced by BASF, ≥98%