Merck
  • [Detecting DNA damage of human lymphocytes exposed to 1,2-DCE with γH2AX identified antibody using flow cytometer assay].
[Detecting DNA damage of human lymphocytes exposed to 1,2-DCE with γH2AX identified antibody using flow cytometer assay].
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases (2011-05-31)
Jun Li, Ying-yan Guo, Wei Wu, Ji-li Bai, Zhi-qiang Xuan, Jun Yang, Jing Wang
ZUSAMMENFASSUNG
To study DNA damage of human peripheral blood lymphocytes exposed to 1,2-dichloroethane (1,2-DCE) with flow cytometry (FCM) assay. The lymphocytes were obtained from 21 workers who are occupationally exposed to 1,2-DCE (exposed group) and 27 workers who were not exposed to 1,2-DCE in the same factory (inner control) and 28 island residents who had never been occupationally exposed to adverse factors (external control). FCM assay was adopted to detect DNA damage of the lymphocytes of each group. Lymphocytes of the health people were incubated with 1,2-DCE at different doses, and FCM assay was used to detect DNA damage. DNA damage rate (%) of the exposed group of exposed workers (4.05% ± 2.55%) was significantly higher than the inner control group of workers (1.97% ± 1.40%) and external control groups of island residents (0.23% ± 0.13%), and the DNA damage of inner control was higher than the external control, all the differences were statistically significant (P < 0.01 or P < 0.05). The geometric mean fluorescence intensity of the workers in the exposed group (3.33 ± 3.01) was significantly higher than the (2.07 ± 0.58) only (P < 0.05). There was no significant difference in the DNA damage rate as well as the geometric mean fluorescence intensity among the exposed group of workers with different years of working period (P > 0.05). In vitro, the fluorescence intensity at the dose of 20, 30 µmol/L for 0.5 h exposure showed statistical significance compared with the negative control group (P < 0.01). The DNA damage rate at the dose of 20, 30 µmol/L for 1.0 h exposure was statistically significant compared with the negative control group (P < 0.05, P < 0.01); The fluorescence intensity at the dose of 10, 20, 30 µmol/L for 1.0 h exposure was statistically significant compared with the negative control group (P < 0.05, P < 0.01). 1,2-DEC can cause DNA damage. And γH2AX FCM assay can be a sensitive, objective and effective method of detecting DNA damage of peripheral blood lymphocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Supelco
1,2-Dichlorethan, analytical standard
Supelco
1,2–Dichlorethan -Lösung, certified reference material, 200 μg/mL in methanol