Merck
  • Design and characterization of a direct ELISA for the detection and quantification of leucomalachite green.
Design and characterization of a direct ELISA for the detection and quantification of leucomalachite green.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2011-05-31)
Gurmit Singh, Terence Koerner, Jean-Marc Gelinas, Michael Abbott, Beth Brady, Anne-Catherine Huet, Caroline Charlier, Philippe Delahaut, Samuel Benrejeb Godefroy
ZUSAMMENFASSUNG
Malachite green (MG), a member of the N-methylated triphenylmethane class of dyes, has long been used to control fungal and protozoan infections in fish. MG is easily absorbed by fish during waterborne exposure and is rapidly metabolized into leucomalachite green (LMG), which is known for its long residence time in edible fish tissue. This paper describes the development of an enzyme-linked immunosorbent assay (ELISA) for the detection and quantification of LMG in fish tissue. This development includes a simple and versatile method for the conversion of LMG to monodesmethyl-LMG, which is then conjugated to bovine serum albumin (BSA) to produce an immunogenic material. Rabbit polyclonal antibodies are generated against this immunogen, purified and used to develop a direct competitive enzyme-linked immunosorbent assay (ELISA) for the screening and quantification of LMG in fish tissue. The assay performed well, with a limit of detection (LOD) and limit of quantification (LOQ) of 0.1 and 0.3 ng g(-1) of fish tissue, respectively. The average extraction efficiency from a matrix of tilapia fillets was approximately 73% and the day-to-day reproducibility for these extractions in the assay was between 5 and 10%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Chlorperbenzoesäure, ≤77%
Sigma-Aldrich
Leucomalachitgrün, powder
Supelco
Leucomalachitgrün, analytical standard