Merck
Multi mycotoxin analysis in food products using immunoaffinity extraction.
Methods in molecular biology (Clifton, N.J.) (2011-06-07)
Masahiko Takino, Hiroki Tanaka, Toshitsugu Tanaka
ZUSAMMENFASSUNG
We developed a method for the simultaneous determination of deoxynivalenol, T-2 toxin, HT-2 toxin and zearalenone in wheat and biscuit by liquid chromatography/electrospray ionization/tandem mass spectrometry coupled with immunoaffinity extraction. This chapter describes a method to extract, clean-up, and quantitate these mycotoxins and the effect of the ion suppression of multifunctional column and IAC in the clean-up were compared.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HT-2-Toxin, ≥98% (HPLC)
Supelco
HT-2 Toxin -Lösung, 100 μg/mL in acetonitrile, analytical standard