Merck
  • Orally active, antimetastatic, nontoxic diphenyl ether-derived carbamoylphosphonate matrix metalloproteinase inhibitors.
Orally active, antimetastatic, nontoxic diphenyl ether-derived carbamoylphosphonate matrix metalloproteinase inhibitors.
ChemMedChem (2011-06-10)
Julia Frant, Ainelly Veerendhar, Tamir Chernilovsky, Shlomo Nedvetzki, Olga Vaksman, Amnon Hoffman, Eli Breuer, Reuven Reich
ZUSAMMENFASSUNG
Seven 4-phenoxybenzenesulfonamidopolymethylene carbamoylphosphonates (CPOs) bearing two to eight methylene units in the polymethylene chain were synthesized and evaluated as matrix metalloproteinase (MMP) inhibitors. The five lowest homologues [(CH₂)₂-₆] are selective MMP-2 inhibitors, whereas the two with the longest linkers [(CH₂)₇,₈] lack inhibitory activity. The most potent homologues are those with (CH₂)₅,₆; these two were evaluated for antimetastatic activity in a murine melanoma model and showed good potency both by oral and intraperitoneal administration without any toxic--including musculoskeletal--side effects. In contrast to the previously reported cis-ACCP, which was shown to inhibit MMP-2 for ∼30 min, the new compounds inhibit MMP activity for the duration of measurement, lasting several hours. Pharmacokinetic evaluation revealed, on the one hand, low oral bioavailability; on the other hand, a relatively large calculated volume of distribution, consistent with the observed reversible absorption of CPO 5 to hydroxyapatite, as a model for bone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diphenylether, ReagentPlus®, ≥99%
Sigma-Aldrich
Diphenylether, ≥99%, FG
Sigma-Aldrich
Diphenylether, ReagentPlus®, 99%
Supelco
Diphenylether, Selectophore, ≥99.9%