Merck
  • Cutting edge: reactive oxygen species inhibitors block priming, but not activation, of the NLRP3 inflammasome.
Cutting edge: reactive oxygen species inhibitors block priming, but not activation, of the NLRP3 inflammasome.
Journal of immunology (Baltimore, Md. : 1950) (2011-06-17)
Franz Bauernfeind, Eva Bartok, Anna Rieger, Luigi Franchi, Gabriel Núñez, Veit Hornung
ZUSAMMENFASSUNG
A common denominator among the multiple damage-inducing agents that ultimately lead to activation of NLRP3 has not yet been identified. Recently, production of reactive oxygen species (ROS) has been suggested to act as a common event upstream of the NLRP3 inflammasome machinery. Because de novo translation of NLRP3 is an essential step in the activation of NLRP3, we investigated the role of substances that inhibit either ROS production or its oxidative activity. Although we observe that NLRP3 inflammasome activation is unique among other known inflammasomes in its sensitivity to ROS inhibition, we have found that this phenomenon is attributable to the fact that NLRP3 strictly requires priming by a proinflammatory signal, a step that is blocked by ROS inhibitors. Although these data do not exclude a general role for ROS production in the process of NLRP3-triggered inflammation, they would put ROS upstream of NLRP3 induction, but not activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diphenyliodonium-hexafluorophosphat, ≥98%
Sigma-Aldrich
Diphenyliodonium-chlorid, ≥98.0% (AT)
Sigma-Aldrich
Diphenyliodoniumnitrat