Merck
Liquid phase separation of polyaromatics on [Cu2(BDC)2(dabco)].
Langmuir : the ACS journal of surfaces and colloids (2011-06-24)
Michael Maes, Stijn Schouteden, Kenji Hirai, Shuhei Furukawa, Susumu Kitagawa, Dirk E De Vos
ZUSAMMENFASSUNG
The porous coordination polymer (PCP) [Cu(2)(BDC)(2)(dabco)] is capable of selectively adsorbing up to 25 wt % of either 1-methylnaphthalene or 2-methylnaphthalene. Uptakes of unsubstituted naphthalene and 1,4-dimethylnaphthalene are significantly lower (7-13 wt %), suggesting that monomethyl substituted polyaromatics can be separated from the other fractions. Furthermore, this PCP can perform the difficult separation of 1-methylnaphthalene from 2-methylnaphthalene with separation factors as high as 2.6, proving that specific interactions of the methyl group with the lattice play an important role in determining the adsorption selectivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Methylnaphthalen, 95%
Sigma-Aldrich
2-Methylnaphthalin (β), 95%
Sigma-Aldrich
1-Methylnaphthalen, ≥95%
Supelco
1-Methylnaphthalen, analytical standard
Supelco
2-Methylnaphthalin, analytical standard