Merck
  • A bilateral comparison study of pimecrolimus cream 1% and a topical medical device cream in the treatment of patients with atopic dermatitis.
A bilateral comparison study of pimecrolimus cream 1% and a topical medical device cream in the treatment of patients with atopic dermatitis.
Journal of drugs in dermatology : JDD (2011-07-02)
Jason J Emer, Amylynne Frankel, Andrew Sohn, Mark Lebwohl
ZUSAMMENFASSUNG
Corticosteroids are the mainstay of therapy for atopic dermatitis, but long-term use is associated with adverse effects. We sought to evaluate the clinical efficacy of two steroid-sparing creams for atopic dermatitis. Twenty patients were enrolled in an investigator-blinded, bilateral comparison study. Patients applied pimecrolimus cream twice daily to a target lesion on one side of the body and also applied a topical medical device cream three times daily on a symmetrical target lesion on the opposite side of the body for four weeks. Clinical assessments including Physician Global Assessment (PGA), Target Lesion Symptom Score (TLSS), subject self-assessment and digital photography were performed at the baseline, 2 week, and 4 week visits. Seventy-five percent of patients (pimecrolimus, 15 of 20; topical medical device, 15 of 20) were rated "clear" (0) or "almost clear" (1) by PGA for both medications after four weeks. Percent improvement of the PGA from randomization for pimecrolimus cream and the topical medical device cream were 72.50 and 71.67 respectively (P=0.9283). PGA scores decreased significantly from baseline for both treatments (P=0.004). Overall, there was no statistically significant difference between treatment groups for PGA scores throughout the study (P=0.8236). No cutaneous side effects were noted. Our study was limited by a small sample size and lack of double-blinding; however, both treatments were found to be safe and effective in treating atopic dermatitis over four weeks. Significant improvements were noted for all efficacy variables. In conclusion, a lipid-rich, non-steroidal, topical medical device cream was as effective in improving atopic dermatitis as pimecrolimus cream.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Brij® 58, average Mn ~1124
Sigma-Aldrich
Brij® C10, average Mn ~683
Sigma-Aldrich
Brij® 52, average Mn ~330
Sigma-Aldrich
ECO BRIJ® C10