Merck
  • Clinical evidence for the role of a topical anti-inflammatory agent in comedonal acne: findings from a randomized study of dapsone gel 5% in combination with tazarotene cream 0.1% in patients with acne vulgaris.
Clinical evidence for the role of a topical anti-inflammatory agent in comedonal acne: findings from a randomized study of dapsone gel 5% in combination with tazarotene cream 0.1% in patients with acne vulgaris.
Journal of drugs in dermatology : JDD (2011-07-02)
Emil Tanghetti, Sunil Dhawan, Lawrence Green, Mark Ling, Jeanine Downie, Marguerite A Germain, J Scott Kasteler, Leon Kircik, Michael G Oefelein, Zoe Draelos
ZUSAMMENFASSUNG
Acne pathogenesis is multifactorial and includes inflammation. Combining drugs targeting multiple components of acne pathogenesis is standard practice. To assess the safety and efficacy of dapsone gel 5%, an anti-inflammatory agent, in combination with tazarotene cream 0.1% for treatment of acne vulgaris. Patients were randomized to receive combination therapy (dapsone gel 5% twice-daily plus tazarotene cream 0.1% daily) or monotherapy (tazarotene cream 0.1% daily). Efficacy and safety data were collected after 1, 2, 4, 8, and 12 weeks of treatment. Patients in both arms (n=86, dapsone + tazarotene; n=85, tazarotene) showed significant reductions from baseline in inflammatory, noninflammatory and total lesion counts (P is less than .001 for all). At 12 weeks, patients treated with dapsone plus tazarotene showed a greater reduction from baseline in noninflammatory (comedonal) and total lesion counts than tazarotene-treated patients (noninflammatory, 59.7 percent vs. 46.5 percent, P=.01; total, 63.3% vs. 53.6%, P=.02). The percentage of patients achieving treatment success (an investigator subjective score of 0 [none] or 1 [minimal]) was greater in dapsone plus tazarotene?treated patients (42.2%) than in tazarotene-treated patients (21.8%;P=.01). Both treatments were well tolerated. Combination therapy with dapsone gel 5% plus tazarotene cream 0.1% was more effective than tazarotene monotherapy for treatment of comedonal acne. The results suggest that anti-inflammatory agents such as dapsone can effectively treat early stages of acne (both comedonal and noncomedonal) when used in combination with a retinoid.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tazarotene, ≥98% (HPLC)