Merck
  • A novel spectrofluorometric method for the determination of methiocarb using an amphiphilic p-sulfonatocalix[4]arene.
A novel spectrofluorometric method for the determination of methiocarb using an amphiphilic p-sulfonatocalix[4]arene.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-07-09)
Xue-Ping Ding, Dong-Bao Tang, Tao Li, Su-Fan Wang, Yun-You Zhou
ZUSAMMENFASSUNG
The characteristics of host-guest complexation between tetrabutyl ether derivatives of p-sulfonatocalix[4]arene (SC4Bu) and methiocarb [3,5-dimethyl-4-(methylthio) phenyl methylcarbamate] were investigated by fluorescence spectrometry. Upon addition of methiocarb, the fluorescence intensity of SC4Bu was quenched regularly and a slight red shift was observed for the maximum emission peak. These results indicated that the SC4Bu-methiocarb complex was formed a 1:1 mole ratio. An association constant of 1.67×10(4) L mol(-1) was calculated by applying a deduced equation. The interaction mechanism of the inclusion complex was discussed. Based on the results, a novel spectrofluorimetric method was described for the determination of methiocarb with a detection limit at 0.05 μg mL(-1). This method is very simple and shows high sensitivity and selectivity. Moreover, the proposed method was successfully applied to the determination of methiocarb in water samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Methiocarb, PESTANAL®, analytical standard