Merck
  • The substrate binding cavity of particulate methane monooxygenase from Methylosinus trichosporium OB3b expresses high enantioselectivity for n-butane and n-pentane oxidation to 2-alcohol.
The substrate binding cavity of particulate methane monooxygenase from Methylosinus trichosporium OB3b expresses high enantioselectivity for n-butane and n-pentane oxidation to 2-alcohol.
Biotechnology letters (2011-07-12)
Akimitsu Miyaji, Teppei Miyoshi, Ken Motokura, Toshihide Baba
ZUSAMMENFASSUNG
The particulate methane monooxygenase (pMMO) of Methylosinus trichosporium OB3b oxidized n-butane and n-pentane and mainly produced (R)-2-butanol and (R)-2-pentanol that comprised 78 and 89% of the product, respectively, indicating that the pro-R hydrogen of the 2-position carbon of n-butane and n-pentane is oriented toward a catalytic site within the substrate binding site of pMMO. The protein cavity adjacent to the catalytic center for pMMO has optimum volume for recognizing n-butane and n-pentane for enantioselective hydroxylation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pentan, ≥99% (GC)
Sigma-Aldrich
2-Butanol, ReagentPlus®, ≥99%
Sigma-Aldrich
Pentan, anhydrous, ≥99%
Sigma-Aldrich
2-Butanol, anhydrous, 99.5%
Sigma-Aldrich
Aluminium-tri-sec-butoxid, 97%
Supelco
2-Butanol, analytical standard
Sigma-Aldrich
(S)-(+)-2-Butanol, 99%
Supelco
Pentan, analytical standard