Merck
Galectin-1 and galectin-8 have redundant roles in promoting plasma cell formation.
Journal of immunology (Baltimore, Md. : 1950) (2011-07-15)
Chih-Ming Tsai, Chin-Huey Guan, Hsiao-Wu Hsieh, Tsui-Ling Hsu, Zhijay Tu, Kuan-Jung Wu, Chun-Hung Lin, Kuo-I Lin
ZUSAMMENFASSUNG
Galectin (Gal) family members are a type of soluble lectin, and they play important roles in immunomodulation. Their redundant roles have been proposed. We previously found that Gal-1 promotes the formation of Ab-secreting plasma cells, but B cells from Gal-1-deficient and control animals produce comparable amounts of Abs. In the current study, we used synthetic sulfomodified N-acetyllactosamine (LacNAc) analogs and short hairpin RNAs for Gal-8 to demonstrate a redundancy in the effects of Gal-1 and Gal-8 on plasma cell formation. Gal-1 and Gal-8 were both expressed during plasma cell differentiation, and both Gals promoted the formation of plasma cells. Gal-1 and Gal-8 bound better to mature B cells than to plasma cells, and the expression of glycosyltransferase enzymes changed during differentiation, with a decrease in mannosyl (α-1,6-)-glycoprotein β-1,6-N-acetyl-glucosaminyltransferase and N-acetylglucosaminyltransferase-1 mRNAs in plasma cells. Synthetic sulfomodified Galβ1-3GlcNAc disaccharides (type 1 LacNAcs) selectively prevented Gal-8 binding, leading to a blockade of Ab production in Gal-1-deficient B cells. Furthermore, synthetic type 1 LacNAcs that were able to block the binding of both Gals greatly reduced the effect of exogenously added recombinant Gal-1 and Gal-8 on promoting Ab production. These results reveal a novel role for Gal-8 in collaboration with Gal-1 in plasma cell formation, and suggest the possibility of using distinct LacNAc ligands to modulate the function of Gals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-D-lactosamin, ≥98% (TLC)