Merck
  • Understanding the dissolution of polyols by ionic liquids using the example of a well-defined model compound.
Understanding the dissolution of polyols by ionic liquids using the example of a well-defined model compound.
Chemphyschem : a European journal of chemical physics and physical chemistry (2011-07-15)
Zakar Papanyan, Christian Roth, Dietmar Paschek, Ralf Ludwig
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pentaerythritol, 99%
Sigma-Aldrich
Pentaerythritol, 98%