Merck
  • Trimethylsilyl trifluoromethanesulfonate-promoted reductive 2'-O-arylmethylation of ribonucleoside derivatives.
Trimethylsilyl trifluoromethanesulfonate-promoted reductive 2'-O-arylmethylation of ribonucleoside derivatives.
Nucleosides, nucleotides & nucleic acids (2011-07-26)
Naoki Uchiyama, Toshihiko Ogata, Natsuhisa Oka, Takeshi Wada
ZUSAMMENFASSUNG
Arylmethyl groups such as benzyl, p-methoxybenzyl, and 1-pyrenylmethyl groups were introduced to the 2'-O-position of nucleosides by reductive etherification. Combining corresponding aromatic aldehydes with 2'-O-trimethylsilylnucleoside derivatives in the presence of trimethylsilyl trifluoromethanesulfonate (TMSOTf) resulted in moderate to good yields of the 2'-O-arylmethyluridine derivatives, whereas the corresponding cytidine and adenosine derivatives were obtained in low yields. The reaction of ribonucleosides with aliphatic aldehydes did not proceed smoothly. Anomerization of the uridine derivatives by TMSOTf was observed in CH(2)Cl(2), toluene, and CH(3)CN, but was completely suppressed when the reactions were conducted in 1,4-dioxane.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trimethylsilyltrifluormethansulfonat, 99%
Sigma-Aldrich
Trimethylsilyltrifluormethansulfonat, purum, ≥98.0% (T)