Merck
  • Adipocyte-derived factors regulate vascular smooth muscle cells through mineralocorticoid and glucocorticoid receptors.
Adipocyte-derived factors regulate vascular smooth muscle cells through mineralocorticoid and glucocorticoid receptors.
Hypertension (Dallas, Tex. : 1979) (2011-07-27)
Aurelie Nguyen Dinh Cat, Ana M Briones, Glaucia E Callera, Alvaro Yogi, Ying He, Augusto C Montezano, Rhian M Touyz
ZUSAMMENFASSUNG
Adipose tissue influences vascular function through adipocyte-derived factors, including components of the renin-angiotensin-aldosterone system. Molecular mechanisms underlying these phenomena are elusive. We investigated the role of adipocyte-derived factors on mitogen-activated protein kinases (MAPKs), proinflammatory status, apoptosis, and mitogenic signaling in vascular smooth muscle cells (VSMCs) and questioned whether these effects involve mineralocorticoid receptor (MR), glucocorticoid receptor (GR), and angiotensin II type 1 receptor (AT(1)R). Cultured mouse VSMCs were exposed to adipocyte-conditioned medium (ACM) from differentiated 3T3-L1 adipocytes. ACM induced phosphorylation of stress-activated protein kinase/c-Jun N-terminal kinase, p38MAPK, and extracellular signal-regulated kinase 1/2 and increased expression of proinflammatory and proliferative markers in VSMCs. Eplerenone (MR antagonist), mifepristone (GR antagonist), and candesartan (AT(1)R antagonist) inhibited ACM-induced effects on extracellular signal-regulated kinase 1/2, p38MAPK, and proliferating cell nuclear antigen, without influencing apoptosis (Bax, Bcl, and caspase 3). Stress-activated protein kinase/c-Jun N-terminal kinase phosphorylation was inhibited by mifepristone and candesartan but not by eplerenone. ACM-induced increase of fibronectin, vascular cell adhesion molecule 1, and plasminogen activator inhibitor 1 expression was blocked by MR and AT(1)R antagonism but not by GR inhibition. ACM has no effect on GR, MR, and AT(1)R expression. Our data show that adipocyte-derived factors influence MAPK signaling, leading to VSMC proinflammatory and profibrotic responses through distinct pathways. Although ACM stimulates p38MAPK and extracellular signal-regulated kinase 1/2 phosphorylation through MR, GR, and AT(1)R, activation of stress-activated protein kinase/c-Jun N-terminal kinase involves GR and AT(1)R. These findings suggest that adipocyte-derived factors regulate VSMC function through specific MAPKs linked to MR, GR, and AT(1)R, a posttranslational phenomenon, because ACM did not influence receptor expression. Such cross-talk between adipocytes and VSMCs may provide a potential molecular mechanism linking renin-angiotensin-aldosterone system, adipocytes, and vascular function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fadrozole hydrochloride, ≥98% (HPLC)