- DPO and POPOP carboxylate-analog sensors by sequential palladium-catalysed direct arylation of oxazole-4-carboxylates.
DPO and POPOP carboxylate-analog sensors by sequential palladium-catalysed direct arylation of oxazole-4-carboxylates.
Organic & biomolecular chemistry (2011-07-29)
Cécile Verrier, Catherine Fiol-Petit, Christophe Hoarau, Francis Marsais
PMID21796283
ZUSAMMENFASSUNG
Sequential palladium-catalysed direct (het)arylation of oxazole-4-carboxylates is achieved to give rapid access to DPO and POPOP (di)carboxylate-analogs. Three novel DPO- and POPOP-type sensors with unusual Stokes shifts and high quantum yields are discovered.