Merck
  • Water increases rates of epoxidation by Mn(III)porphyrins/imidazole/IO4(-) in CH2Cl2. Analogy with peroxidase and chlorite dismutase.
Water increases rates of epoxidation by Mn(III)porphyrins/imidazole/IO4(-) in CH2Cl2. Analogy with peroxidase and chlorite dismutase.
Dalton transactions (Cambridge, England : 2003) (2011-08-02)
Leila Mahmoudi, Daryoush Mohajer, Reinhard Kissner, Willem H Koppenol
ZUSAMMENFASSUNG
Manganese(III)-meso-tetraphenylporphyrin [Mn(TPP)] and manganese(III)-meso-tetrakis(pentafluorophenyl)porphyrin [Mn(TPFPP)] catalyse the epoxidation of cyclooctene by IO(4)(-) in the presence of excess imidazoles, in both dry CH(2)Cl(2) and CH(2)Cl(2) saturated with H(2)O. The reaction rates of the electron deficient Mn(TPFPP) are a factor 24 less than those of Mn(TPP); however, the former increases 15-30 times in the presence of water, while those of Mn(TPP) do so by a factor of 2-3. The most striking catalytic enhancement caused by the addition of water was observed with 2-methylimidazole and Mn(TPFPP). As deprotonation of imidazoles may play a significant role in the presence of water, we found that manganese(III)-meso-tetrakis(phenyl-4-sulfonato)porphyrin [Mn(TPPS)] decreases the NH proton pK(a) of axially coordinated imidazole from 14.2 to 9.5. We conclude that the imidazole ligand is partially deprotonated in the presence of water. The latter enables the solvation of imidazolium ions that are formed simultaneously. The imidazolate form of the co-catalyst is a much stronger donor than the imidazole itself, providing electron density to Mn(III) and thus promoting oxygen transfer. The failure of N-methylimidazole to increase the reaction rates upon addition of water supports this hypothesis. A functionally related deprotonation has been shown to occur in horseradish peroxidase (J. S. de Ropp, V. Thanabal, G. N. La Mar, J. Am. Chem. Soc. 1985, 107, 8270-8272) and in chlorite dismutase (B. R. Goblirsch, B. R. Streit, J. L. Dubois, C. M. Wilmot, J. Biol. Inorg. Chem. 2010, 15, 879-888). Mn(III)porphyrins in combination with imidazoles and water constitute a functional biomimetic model of peroxidases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Supelco
1,2-Dichlorethan, analytical standard
Supelco
1,2–Dichlorethan -Lösung, certified reference material, 200 μg/mL in methanol