Merck
  • Cloning of a novel glutathione S-transferase 3 (GST3) gene and expressionanalysis in pearl oyster, Pinctada martensii.
Cloning of a novel glutathione S-transferase 3 (GST3) gene and expressionanalysis in pearl oyster, Pinctada martensii.
Fish & shellfish immunology (2011-08-03)
Jinhui Chen, Shu Xiao, Yuewen Deng, Xiaodong Du, Ziniu Yu
ZUSAMMENFASSUNG
Microsomal glutathione S-transferase (MGST) functions in cellular defense against xenobiotics and provides protection against the action of lipid hydroperoxides produced as a consequence of oxidative stress. In this study, a full-length cDNA encoding MGST3 (referred to as PmMGST3) was identified from the pearl oyster, Pinctada martensii by a combination of expressed sequence tag (EST) analysis and rapid amplification of cDNA ends (RACE). The full-length cDNA of PmMGST3 is 971 bp and contains a 5' UTR of 39 bp, a 3' UTR of 491 bp with a canonical polyadenylation signal sequence (AATAAA), and an open reading frame (ORF) of 447 bp encoding a polypeptide of 146 residues. The deduced polypeptide contains a conserved motif (FNCx(1)QRx(2)H) characteristic of the MGST3 subfamily. The PmMGST3 transcript could be detected in all tissues tested, with highest transcript level seen in hepatopancreas. Cadmium treatment significantly increased PmMGST3 mRNA levels in gill and hepatopancreas, while bacterial challenge initially depressed mRNA levels and then increased its level in haemocytes, gill and hepatopancreas in a time-dependent manner. In an assay using cumene hydroperoxide as a substrate, we demonstrated that PmMGST3 possesses glutathione-dependent peroxidase activity. These results suggest that PmMGST3 plays an important role in cellular defense against oxidative stress caused by cadmium and bacteria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cumolhydroperoxid, technical grade, 80%