Merck
  • Preparation and evaluation of molecularly imprinted ionic liquids polymer as sorbent for on-line solid-phase extraction of chlorsulfuron in environmental water samples.
Preparation and evaluation of molecularly imprinted ionic liquids polymer as sorbent for on-line solid-phase extraction of chlorsulfuron in environmental water samples.
Journal of chromatography. A (2011-08-03)
Lu Guo, Qiliang Deng, Guozhen Fang, Wei Gao, Shuo Wang
ZUSAMMENFASSUNG
In this report, vinylimidazolium ionic liquid as a functional monomer for preparation of chlorsulfuron (CS) imprinted polymers were first studied. The imprinted materials showed high selectivity for CS, and fast kinetics so that adsorption equilibrium was achieved within 5 min. These materials have been further employed to detect trace CS from water samples by online preconcentration coupled with HPLC. The sorbent offered good linearity (0.005-30 μg L(-1), r(2)>0.99) for on-line solid-phase extraction of trace chlorsulfuron. Under the optimal experimental conditions, the recovery for chlorsulfuron was in the range of 81.0-110.1% for the water samples, with RSDs ranging from 1.2 to 7.6%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Chlorsulfuron, PESTANAL®, analytical standard