Merck
Acquired alpha 1-antitrypsin deficiency in tropical pulmonary eosinophilia.
The Indian journal of medical research (2011-08-03)
Debidas Ray, S Harikrishna, Chandra Immanuel, Lalitha Victor, Sudha Subramanyam, V Kumaraswami
ZUSAMMENFASSUNG
Observation of an increased frequency of an intermediate deficiency of serum alpha1-antitrypsin (α1-AT) in patients with Tropical Pulmonary Eosinophilia (TPE) was earlier reported. Though the possibility of existence of an acquired deficiency was suggested, without phenotyping a hereditary α1-AT deficiency in TPE could not totally be ruled out. In this study, we have done Pi (Protease inhibitor) phenotyping to investigate the possibility of association of any heterozygous (or homozygous) α1-AT deficiency in patients with TPE. Serum a1antitrypsin (α1-AT) was measured in 103 patients (Group A) with TPE, 99 patients with pulmonary eosinophilia who had associated intestinal worm infestation (Group B) and 43 healthy volunteers who served as controls. In 19 α1-AT deficient patients (9 of Group A and 10 of Group B), α1-AT level was measured before and after treatment. In 58 patients with TPE and in 5 controls, phenotyping was done. Fifteen patients of Group A and 16 from Group B showed intermediate α1-AT deficiency (150 mg % or less. None of the control subjects had α1-AT deficiency (<200 mg%). After treatment with DEC and/or deworming, in 19 patients there was a significant (P < 0.001) rise in α1-AT levels. Results of phenotyping showed that all had M1 or M 2 allele and none had S or Z variant (either homozygous or heterozygous) thus ruling out any underlying genetic cause for the observed α1-AT deficiency. The observed α1-AT deficiency may be due to the chronic inflammation in TPE and associated oxidative stress. However, in such α1-AT deficient patients with TPE and those with worm infested pulmonary eosinophilia, faecal α1-AT concentration and faecal α1-AT clearance should be routinely estimated to rule out the possibility of any intestinal protein loss.